siRNA Details
Gene Symbol: Ppp2r3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163415.1 | NP_001156887.1 | 1189 | |||
NM_011154.2 | NP_035284.2 | 955 | ||||
GenBank mRNA | AF050165.1 | - | 952 | |||
AK054122.1 | - | 1328 | ||||
AK143212.1 | - | 1232 | ||||
BC027415.1 | - | 932 | 4235453 | |||
BC059852.1 | - | 1220 | 6402777 | |||
BC095938.1 | - | 110 | ||||
BC096544.1 | - | 635 | 6834351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
