siRNA Details
Gene Symbol: Psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011187.1 | NP_035317.1 | 4, 5 | 407 | ||
GenBank mRNA | AK002823.1 | - | 4, 5 | 422 | ||
AK012613.1 | - | 4, 5 | 408 | |||
AK013961.1 | - | 4, 5 | 418 | |||
AK031913.1 | - | 4, 5 | 423 | |||
AK075759.1 | - | 4, 5 | 423 | |||
AK088276.1 | - | 4, 5 | 418 | |||
AK088765.1 | - | 4, 5 | 421 | |||
AK162002.1 | - | 4, 5 | 426 | |||
AK168823.1 | - | 4, 5 | 410 | |||
BC033424.1 | - | 4, 5 | 404 | |||
BC049230.1 | - | 4, 5 | 400 | |||
BC057662.1 | - | 4, 5 | 408 | 2865594 | ||
D83585.1 | - | 4, 5 | 407 | |||
Y10874.1 | - | 4, 5 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
