siRNA Details
Gene Symbol: Rpl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011291.5 | NP_035421.2 | 2 | 230 | ||
GenBank mRNA | AK017074.1 | - | 2 | 200 | ||
AK050656.1 | - | 2 | 205 | |||
AK088291.1 | - | 2 | 204 | |||
AK135460.1 | - | 2 | 200 | |||
AK146219.1 | - | 2 | 201 | |||
AK167124.1 | - | 2 | 204 | |||
AK168137.1 | - | 2 | 200 | |||
AK168227.1 | - | 2 | 200 | |||
BB839528.1 | - | 2 | 230 | |||
BC025909.1 | - | 2 | 180 | 5027545 | ||
BC051261.1 | - | 2 | 196 | 30072081 | ||
BC086786.1 | - | 1 | 176 | 5701071 | ||
BC096452.1 | - | 2 | 183 | 3588544 | ||
M29016.1 | - | 2 | 199 | |||
M85235.1 | - | 2 | 185 |
Gene Symbol: 1700073E17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC049631.1 | - | 2 | 382 |

More Information
Other Species Targeted

Gene Ontology Categories:
