siRNA Details
Gene Symbol: Rfxank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025589.1 | NP_001020760.1 | 9 | 1834 | ||
NM_011266.2 | NP_035396.1 | 9 | 1864 | |||
GenBank mRNA | AF094761.1 | - | 10 | 1336 | ||
AK052298.1 | - | 9 | 1791 | |||
AK087462.1 | - | 9 | 1765 | |||
AK162595.1 | - | 5 | 1452 | |||
BC010971.1 | - | 9 | 1770 | 4190691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
