siRNA Details
Gene Symbol: Saa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011314.2 | NP_035444.1 | 4 | 590 | ||
GenBank mRNA | BC034858.1 | - | 1 | 2164 | ||
M11130.1 | - | 4 | 589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
