siRNA Details
Gene Symbol: Shbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011367.2 | NP_035497.1 | 8 | 1285 | ||
GenBank mRNA | AK010959.1 | - | 8 | 1284 | ||
BC120785.1 | - | 8 | 1270 | 40129708 | ||
BC120787.1 | - | 8 | 1270 | 40129710 | ||
U85644.1 | - | 8 | 1248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
