siRNA Details
Gene Symbol: Serpinb9g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011455.3 | NP_035585.2 | 4 | 397 | ||
GenBank mRNA | AF425083.1 | - | 4 | 392 | ||
AK005394.1 | - | 4 | 397 | |||
BC064758.1 | - | 4 | 382 | 30018322 | ||
BC064759.1 | - | 4 | 372 | 30432705 |
Gene Symbol: Serpinb9d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011460.2 | NP_035590.1 | 4 | 397 | ||
GenBank mRNA | AK028382.1 | - | 4 | 398 | ||
BC100308.1 | - | 3 | 348 | |||
BC139261.1 | - | 4 | 373 | 8862283 | ||
BC139263.1 | - | 4 | 373 | 8862285 |
Gene Symbol: Serpinb9f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183197.1 | NP_899020.1 | 4 | 386 | ||
GenBank mRNA | BC052216.1 | - | 4 | 386 | 30020424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
