siRNA Details
Gene Symbol: Slpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011414.3 | NP_035544.1 | 1, 2 | 319 | ||
GenBank mRNA | BC028509.1 | - | 1, 2 | 319 | 1513866 | |
U73004.1 | - | 530 | ||||
U88093.1 | - | 1, 2 | 96 | |||
U94341.1 | - | 1, 2 | 97 |

More Information
Other Species Targeted

Gene Ontology Categories:
