siRNA Details
Gene Symbol: CACNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000726.3 | NP_000717.2 | 2 | 165 | ||
NM_001005746.2 | NP_001005746.1 | 2 | 115 | |||
NM_001145798.1 | NP_001139270.1 | 2 | 165 | |||
XM_011511797.2 | XP_011510099.1 | 2 | 1074 | |||
XM_011511798.2 | XP_011510100.1 | 2 | 1074 | |||
XM_017004880.1 | XP_016860369.1 | 2 | 166 | |||
XM_017004887.1 | XP_016860376.1 | 2 | 1074 | |||
XM_017004888.1 | XP_016860377.1 | 2 | 174 | |||
GenBank mRNA | AB302276.1 | - | 2 | 109 | ||
AF038852.1 | - | 2 | 75 | |||
AK291378.1 | - | 3 | 248 | |||
AK294398.1 | - | 2 | 89 | |||
AK316045.1 | - | 2 | 106 | |||
BC075049.2 | - | 2 | 116 | 30915578 | ||
BP360802.1 | - | 2 | 98 | |||
BX279713.1 | - | 2 | 115 | |||
U95020.1 | - | 2 | 165 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001285426, NM_001037099, NM_001285428, AK032311, AK158606, and others.
Gene Ontology Categories:
