siRNA Details
Gene Symbol: Strn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011500.2 | NP_035630.2 | 2 | 344 | ||
GenBank mRNA | AJ223777.1 | - | 2 | 271 | ||
AK051549.1 | - | 2 | 346 | |||
BC090968.1 | - | 2 | 344 | 30646517 | ||
BC150727.1 | - | 2 | 312 | 9087638 |
