siRNA Details
Gene Symbol: Slc7a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253679.1 | NP_001240608.1 | 11 | 1909 | ||
NM_001253680.1 | NP_001240609.1 | 10 | 1927 | |||
NM_011405.4 | NP_035535.2 | 12 | 2066 | |||
GenBank mRNA | AJ012754.1 | - | 10 | 1917 | ||
AJ130943.1 | - | 11 | 1909 | |||
AK150950.1 | - | 12 | 2066 | |||
AK151718.1 | - | 12 | 2066 | |||
AK152637.1 | - | 10 | 1832 | |||
AK170851.1 | - | 10 | 1838 | |||
BC014709.1 | - | 10 | 1915 | 4482816 |

More Information
Other Species Targeted

Gene Ontology Categories:
