siRNA Details
Gene Symbol: Med22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011513.2 | NP_035643.1 | 3 | 2289 | ||
GenBank mRNA | AK144744.1 | - | 3 | 2268 | ||
AK161053.1 | - | 3 | 2230 | |||
AK166250.1 | - | 3 | 2238 | |||
AK166474.1 | - | 3 | 2262 | |||
AK198745.1 | - | 1 | 80 | |||
BC018225.1 | - | 3 | 2279 | 3968506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
