siRNA Details
Gene Symbol: Tbx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009323.2 | NP_033349.2 | 8 | 1428 | ||
GenBank mRNA | AK132578.1 | - | 8 | 1429 | ||
AK137458.1 | - | 8 | 1222 | |||
AY662679.1 | - | 8 | 1361 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008761285, XM_006233027.
Gene Ontology Categories:
