siRNA Details
Gene Symbol: Skp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011543.4 | NP_035673.3 | 3 | 250 | ||
GenBank mRNA | AF083214.1 | - | 3 | 174 | ||
AK002983.1 | - | 3 | 221 | |||
AK010628.1 | - | 3 | 236 | |||
AK012498.1 | - | 3 | 209 | |||
AK014245.1 | - | 3 | 209 | |||
AK027909.1 | - | 3 | 208 | |||
AK088339.1 | - | 3 | 209 | |||
AK152609.1 | - | 3 | 208 | |||
AK160198.1 | - | 3 | 209 | |||
AK166673.1 | - | 3 | 209 | |||
BC002115.1 | - | 3 | 193 | 3490159 | ||
BU841654.1 | - | 4 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
