siRNA Details
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145234.1 | - | 2 | 511 | ||
KJ650229.1 | - | 1 | 401 | |||
U60087.1 | - | 1 | 401 | |||
U60088.1 | - | 1 | 401 | |||
U60089.1 | - | 1 | 401 | |||
U60090.1 | - | 1 | 401 | |||
U60091.1 | - | 1 | 401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
