siRNA Details
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161722.1 | NP_001155194.1 | 3 | 580 | ||
NM_001161723.1 | NP_001155195.1 | 3 | 574 | |||
NM_011549.3 | NP_035679.3 | 2 | 521 | |||
GenBank mRNA | AF079095.1 | - | 2 | 502 | ||
AK078755.1 | - | 3 | 512 | |||
AK146016.1 | - | 2 | 546 | |||
AK154971.1 | - | 3 | 578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
