siRNA Details
Gene Symbol: Alyref | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011568.1 | NP_035698.1 | 5, 6 | 787 | ||
GenBank mRNA | AK035721.1 | - | 195 | |||
BC120588.1 | - | 5, 6 | 879 | 40129511 | ||
BC137658.1 | - | 5, 6 | 879 | 8860677 | ||
U89876.1 | - | 5, 6 | 787 |

More Information
Other Species Targeted

Gene Ontology Categories:
