siRNA Details
Gene Symbol: Try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038996.2 | NP_001034085.1 | 5 | 764 | ||
GenBank mRNA | AK133516.1 | - | 5 | 691 | ||
BC138698.1 | - | 5 | 764 | 8861720 | ||
BC146017.1 | - | 5 | 764 | 40131320 |
Gene Symbol: Gm5409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003664.2 | NP_001003664.1 | 5 | 680 |
Gene Symbol: Try4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011646.5 | NP_035776.1 | 5 | 690 | ||
GenBank mRNA | AK003064.1 | - | 5 | 694 | ||
AK008660.1 | - | 5 | 691 | |||
AK008667.1 | - | 5 | 691 | |||
BC050120.1 | - | 5 | 671 | |||
BC061135.1 | - | 5 | 690 | 30306436 |
Gene Symbol: Try5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003405.4 | NP_001003405.1 | 5 | 692 | ||
GenBank mRNA | AA168368.1 | - | 2 | 148 | ||
AB009661.1 | - | 5 | 692 | |||
AK007843.1 | - | 5 | 691 | |||
AK131898.1 | - | 5 | 691 | |||
AK160484.1 | - | 5 | 691 | |||
BC139220.1 | - | 5 | 777 | 8862242 | ||
BC139223.1 | - | 5 | 777 | 8862245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
