siRNA Details
Gene Symbol: Alyref | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011568.1 | NP_035698.1 | 6 | 1101 | ||
GenBank mRNA | AK035721.1 | - | 509 | |||
U89876.1 | - | 6 | 1101 |
Gene Symbol: Alyref2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019484.4 | NP_062357.3 | 1 | 1098 | ||
GenBank mRNA | AK048243.1 | - | 1 | 1098 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
