siRNA Details
Gene Symbol: Cd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ401387.1 | - | 7, 8 | 621 | ||
AJ401388.1 | - | 7, 8 | 784 | |||
AJ401389.1 | - | 8, 9 | 678 | |||
AJ401390.1 | - | 7, 8 | 596 | |||
AK089861.1 | - | 702 | ||||
AK156644.1 | - | 8, 9 | 717 | |||
M83312.1 | - | 8, 9 | 685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
