siRNA Details
Gene Symbol: Xlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291747.1 | NP_001278676.1 | 5 | 276 | ||
NM_001291748.1 | NP_001278677.1 | 5 | 284 | |||
NM_011725.4 | NP_035855.1 | 5 | 324 | |||
GenBank mRNA | AA623498.1 | - | 5 | 238 | ||
AK012549.1 | - | 5 | 315 | |||
AK035025.1 | - | 5 | 286 | |||
AK035341.1 | - | 5 | 293 | |||
AK088930.1 | - | 5 | 228 | |||
BC005560.1 | - | 5 | 311 | 3965533 | ||
X07967.1 | - | 5 | 259 |
Gene Symbol: 3830403N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014417.1 | - | 4 | 304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
