siRNA Details
Gene Symbol: Zfp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011747.2 | NP_035877.1 | 3 | 556 | ||
GenBank mRNA | AK030240.1 | - | 3 | 557 | ||
BC052082.1 | - | 3 | 459 | 6404424 | ||
FJ593171.1 | - | 3 | 492 |
