siRNA Details
Gene Symbol: Xirp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF051945.2 | - | 2 | 3219 | ||
BC117959.1 | - | 2 | 3153 | 40098186 | ||
BC117960.1 | - | 2 | 3153 | 40098187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
