siRNA Details
Gene Symbol: Coro1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011778.2 | NP_035908.1 | 11 | 1726 | ||
GenBank mRNA | AF143956.1 | - | 11 | 1733 | ||
AK008947.1 | - | 7 | 1166 | |||
AK149639.1 | - | 11 | 1738 | |||
AK159329.1 | - | 9 | 1418 | |||
AK185002.1 | - | 1 | 219 | |||
AK213696.1 | - | 1 | 219 | |||
BC029326.1 | - | 11 | 1685 | |||
BC035545.1 | - | 1 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
