siRNA Details
Gene Symbol: Dmtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110327.1 | NP_001103797.1 | 4 | 458 | ||
NM_011806.3 | NP_035936.3 | 4 | 458 | |||
GenBank mRNA | AK029147.1 | - | 4 | 400 | ||
AK030508.1 | - | 4 | 427 | |||
AK033208.1 | - | 4 | 424 | |||
AK039563.1 | - | 4 | 426 | |||
AK040107.1 | - | 4 | 416 | |||
AK046201.1 | - | 4 | 354 | |||
AK078015.1 | - | 4 | 359 | |||
AK145803.1 | - | 4 | 420 | |||
AK160595.1 | - | 4 | 431 | |||
BC060109.1 | - | 4 | 381 | |||
U70017.1 | - | 4 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
