siRNA Details
Gene Symbol: Fbxl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015794.1 | NP_056609.1 | 9 | 9743 | ||
GenBank mRNA | AK018254.1 | - | 1 | 787 | ||
AK086279.1 | - | 1 | 1620 | |||
AK160716.1 | - | 1 | 381 |
Gene Symbol: 5530400K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017425.1 | - | 1 | 1113 |
Gene Symbol: Oas1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB067529.1 | - | 6 | 1687 | ||
AF418004.1 | - | 6 | 1688 | |||
AK080152.1 | - | 6 | 1681 |

More Information
Other Species Targeted

Gene Ontology Categories:
