siRNA Details
Gene Symbol: C1ql1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011795.2 | NP_035925.2 | 2 | 695 | ||
GenBank mRNA | AF095155.1 | - | 2 | 695 | ||
BC022724.1 | - | 2 | 282 | |||
BC118980.2 | - | 2 | 695 | 40100122 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108838.
Gene Ontology Categories:
