siRNA Details
Gene Symbol: Adam25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011781.2 | NP_035911.2 | 2 | 1378 | ||
GenBank mRNA | AF167403.1 | - | 1378 | |||
AK016581.1 | - | 2 | 1376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
