siRNA Details
Gene Symbol: Mrps12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011885.4 | NP_036015.1 | 3 | 575 | ||
GenBank mRNA | AK146730.1 | - | 3 | 598 | ||
BC027522.1 | - | 3 | 580 | 5058479 | ||
Y11682.1 | - | 3 | 574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
