siRNA Details
Gene Symbol: Psg19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011964.2 | NP_036094.2 | 3 | 769 | ||
GenBank mRNA | BC099467.1 | - | 3 | 745 | 30713077 | |
X98111.1 | - | 3 | 769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
