siRNA Details
Gene Symbol: GRIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000828.4 | NP_000819.3 | 4 | 873 | ||
NM_007325.4 | NP_015564.4 | 4 | 873 | |||
GenBank mRNA | AK297136.1 | - | 4 | 559 | ||
BC117464.1 | - | 4 | 745 | 40126015 | ||
U10301.1 | - | 4 | 633 | |||
U10302.1 | - | 4 | 633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
