siRNA Details
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039072.2 | NP_001034161.1 | 4, 5 | 436 | ||
NM_001039073.2 | NP_001034162.1 | 4, 5 | 436 | |||
NM_001039074.2 | NP_001034163.1 | 3, 4 | 467 | |||
NM_001039075.2 | NP_001034164.1 | 4, 5 | 436 | |||
GenBank mRNA | AF114379.2 | - | 4, 5 | 400 | ||
AJ005621.1 | - | 4, 5 | 429 | |||
AK004020.1 | - | 4, 5 | 444 | |||
AK172980.1 | - | 3, 4 | 467 | |||
AY206013.1 | - | 4, 5 | 400 | |||
AY206015.1 | - | 3, 4 | 352 | |||
BC099596.1 | - | 4, 5 | 403 | |||
BY704269.1 | - | 4, 5 | 443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
