siRNA Details
Gene Symbol: Cln8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012000.3 | NP_036130.1 | 3 | 1280 | ||
GenBank mRNA | AF125307.1 | - | 3 | 770 | ||
AF125308.1 | - | 3 | 770 | |||
AK049145.1 | - | 3 | 953 | |||
AK080049.1 | - | 3 | 788 | |||
AK088300.1 | - | 3 | 934 | |||
AK089692.1 | - | 3 | 929 | |||
AK128890.1 | - | 3 | 951 | |||
AK164268.1 | - | 3 | 953 | |||
AK166905.1 | - | 3 | 1705 | |||
AK169698.1 | - | 3 | 934 | |||
AK171446.1 | - | 3 | 962 | |||
AK201598.1 | - | 1 | 17 | |||
BC021625.1 | - | 3 | 786 | 4188527 |
