siRNA Details
Gene Symbol: ASIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001094.4 | NP_001085.2 | 1 | 702 | ||
GenBank mRNA | AK313043.1 | - | 1 | 671 | ||
BC075042.2 | - | 1 | 469 | 30915358 | ||
BC075043.2 | - | 1 | 469 | 30915574 | ||
U50352.1 | - | 1 | 428 | |||
U53212.1 | - | 1 | 555 | |||
U57352.1 | - | 1 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
