siRNA Details
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290635.1 | NP_001277564.1 | 4 | 605 | ||
NM_012018.2 | NP_036148.2 | 14 | 2247 | |||
GenBank mRNA | AK088733.1 | - | 4 | 607 | ||
BC050831.1 | - | 6 | 879 | |||
BC060146.1 | - | 4 | 581 | |||
BC079536.1 | - | 2 | 269 | |||
BC158010.1 | - | 13 | 2120 | 9007318 | ||
X79131.1 | - | 6 | 907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
