siRNA Details
Gene Symbol: Anxa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087941.1 | - | 4 | 290 | ||
AK169774.1 | - | 4 | 290 | |||
BC005595.1 | - | 4 | 256 | 3482035 | ||
X13460.1 | - | 3 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
