siRNA Details
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271496.1 | NP_001258425.1 | 4 | 868 | ||
NM_013490.4 | NP_038518.2 | 3 | 814 | |||
GenBank mRNA | AB011002.1 | - | 3 | 673 | ||
AK005057.1 | - | 5 | 929 | |||
AK166465.1 | - | 3 | 755 | |||
AK178697.1 | - | 2 | 84 | |||
AK183570.1 | - | 2 | 88 | |||
AK186560.1 | - | 2 | 88 | |||
AK192596.1 | - | 2 | 84 | |||
AK215104.1 | - | 3 | 207 | |||
BC060218.1 | - | 3 | 777 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017127, XM_008760090, XM_006230708, XM_006230707, XM_008760091, and others.
Gene Ontology Categories:
