siRNA Details
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012003.2 | NP_036133.1 | 8 | 1636 | ||
GenBank mRNA | AF071316.1 | - | 8 | 1638 | ||
BC003724.1 | - | 7 | 1517 | 3593979 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
