siRNA Details
Gene Symbol: Srp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012058.3 | NP_036188.1 | 3 | 508 | ||
GenBank mRNA | AK011720.1 | - | 3 | 436 | ||
AK020583.1 | - | 3 | 457 | |||
AK051208.1 | - | 3 | 454 | |||
AK146882.1 | - | 3 | 457 | |||
AK169314.1 | - | 3 | 456 | |||
CJ047061.1 | - | 2 | 440 | |||
X78304.1 | - | 3 | 434 | |||
X78305.1 | - | 1 | 842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
