siRNA Details
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271496.1 | NP_001258425.1 | 5 | 1031 | ||
NM_013490.4 | NP_038518.2 | 4 | 977 | |||
GenBank mRNA | AB011002.1 | - | 4 | 836 | ||
AK005057.1 | - | 6 | 1092 | |||
AK166465.1 | - | 4 | 918 | |||
AK216139.1 | - | 1 | 83 | |||
BC004719.1 | - | 2 | 167 | |||
BC060218.1 | - | 4 | 940 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017127, XM_008760090, XM_006230708, XM_006230707, XM_008760091, and others.
Gene Ontology Categories:
