siRNA Details
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083453.1 | - | 1 | 1675 | ||
AK206779.1 | - | 6 | 188 | |||
AK219294.1 | - | 6 | 188 | |||
BC055883.1 | - | 4 | 374 | |||
BC059726.1 | - | 6 | 202 | |||
BC080708.1 | - | 2 | 921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
