siRNA Details
Gene Symbol: H2-M10.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013544.3 | NP_038572.3 | 2 | 152 | ||
GenBank mRNA | AF539597.1 | - | 2 | 152 | ||
BC104321.1 | - | 2 | 144 | |||
BC119458.2 | - | 2 | 152 | 40044798 | ||
BC152395.1 | - | 2 | 144 |
Gene Symbol: H2-M10.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177634.1 | NP_808302.1 | 2 | 152 | ||
GenBank mRNA | AF539600.1 | - | 2 | 152 | ||
AY211081.1 | - | 2 | 140 |

More Information
Other Species Targeted

Gene Ontology Categories:
