siRNA Details
Gene Symbol: Dmtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252662.1 | NP_001239591.1 | 4 | 640 | ||
NM_001252663.1 | NP_001239592.1 | 4 | 640 | |||
NM_001252664.1 | NP_001239593.1 | 3 | 467 | |||
NM_001252665.1 | NP_001239594.1 | 3 | 366 | |||
NM_001252666.1 | NP_001239595.1 | 2 | 399 | |||
NM_013514.4 | NP_038542.1 | 5 | 581 | |||
GenBank mRNA | AF079846.3 | - | 3 | 310 | ||
AF155547.1 | - | 3 | 310 | |||
AK046217.1 | - | 1 | 365 | |||
AK143840.1 | - | 4 | 632 | |||
AK149301.1 | - | 3 | 277 | |||
AK155927.1 | - | 4 | 634 | |||
AK158744.1 | - | 3 | 398 | |||
AK162866.1 | - | 3 | 318 | |||
AK165204.1 | - | 633 | ||||
AK165294.1 | - | 4 | 632 | |||
BC016897.1 | - | 5 | 581 | 4240747 | ||
BC037021.1 | - | 2 | 386 | 5366040 | ||
BY270342.1 | - | 3 | 367 |

More Information
Other Species Targeted

Gene Ontology Categories:
