siRNA Details
Gene Symbol: H2-M10.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013544.3 | NP_038572.3 | 2 | 182 | ||
GenBank mRNA | AF539597.1 | - | 2 | 182 | ||
AY211079.1 | - | 2 | 182 | |||
BC104321.1 | - | 2 | 174 | |||
BC119458.2 | - | 2 | 182 | 40044798 | ||
BC152395.1 | - | 2 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
