siRNA Details
Gene Symbol: Mbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF072248.1 | - | 3, 4 | 660 | ||
BC038264.1 | - | 3, 4 | 461 | 5250237 | ||
BQ042794.1 | - | 2, 3 | 228 | |||
CK625083.1 | - | 3, 4 | 480 |

More Information
Other Species Targeted

