siRNA Details
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083453.1 | - | 4 | 2496 | ||
BC055883.1 | - | 8 | 1006 | |||
BC059726.1 | - | 8 | 284 | |||
BC080708.1 | - | 6 | 1552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
