siRNA Details
Gene Symbol: Hebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013546.2 | NP_038574.2 | 4 | 971 | ||
GenBank mRNA | AB013095.1 | - | 4 | 910 | ||
AF117613.1 | - | 4 | 959 | |||
AK008919.1 | - | 4 | 892 | |||
BC012654.1 | - | 4 | 896 | 4216585 | ||
CK378928.1 | - | 1 | 226 |
