siRNA Details
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113199.1 | NP_001106670.1 | 3 | 2566 | ||
GenBank mRNA | U48364.1 | - | 3 | 2277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
