siRNA Details
Gene Symbol: Dusp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013642.3 | NP_038670.1 | 3 | 802 | ||
GenBank mRNA | AK145903.1 | - | 3 | 807 | ||
AK149915.1 | - | 3 | 680 | |||
AK150433.1 | - | 3 | 802 | |||
AK152183.1 | - | 3 | 802 | |||
AK152324.1 | - | 3 | 805 | |||
AK152905.1 | - | 2 | 1172 | |||
AK159251.1 | - | 3 | 803 | |||
AK159363.1 | - | 3 | 803 | |||
AK159567.1 | - | 3 | 803 | |||
AK159658.1 | - | 3 | 800 | |||
AK159726.1 | - | 3 | 736 | |||
AK167602.1 | - | 3 | 804 | |||
AK168627.1 | - | 3 | 803 | |||
BC006967.1 | - | 3 | 789 | 3589849 | ||
X61940.1 | - | 3 | 792 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053769, U02553, S81478, X84004.
Gene Ontology Categories:
