siRNA Details
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013648.6 | NP_038676.1 | 2, 3 | 295 | ||
GenBank mRNA | AF038538.1 | - | 2, 3 | 251 | ||
AK144271.1 | - | 2, 3 | 258 | |||
AK165915.1 | - | 2, 3 | 285 | |||
AU066588.1 | - | 2, 3 | 296 | |||
BC031370.1 | - | 2, 3 | 252 | 5359358 | ||
CJ176292.1 | - | 2, 3 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
